GsBP®-FFAP Polar, PEG GC Columns, GS-Tek
Supplier: General Separation Technologies, Inc.
2120-2503 2120-5003 2125-1502 2125-3002 2125-3005 2125-6002 2125-6005 2132-1502 2132-2505 2132-3002 2132-3005 2132-3010 2132-5005 2132-6005 2132-6010 2153-1010 2153-1505 2153-1510 2153-3002 2153-3005 2153-3010 2153-3015 2153-6010
10499-500EA 1365.12 CAD
10499-500 10499-502 10499-504 10499-506 10499-524 10499-526 10499-528 10499-530 10499-532 10499-534 10499-536 10499-538 10499-540 10499-542 10499-544 10499-546 10499-548 10499-550 10499-552 10499-554 10499-556 10499-558 10499-560
GsBP®-FFAP Polar, PEG GC Columns, GS-Tek
Chromatography Columns
Typical Application: Acrylates, Alcoholic Beverage Standard: Acids & Esters, Aspirin and Ibuprofen in Methanol, Cresylic Acids, Ethoxyethanol, Fatty Acids (Free), Malt Whiskey, Organic Acids.
  • Bondable polyethylene glycol
  • Acid modified
  • High polar
  • Equivalent to USP G25 and G35
Ordering information: These columns are suitable replacements for HP-FFAP, DB-FFAP, Rxt-Stabilwa-DA, CP-FFAP.