Single Specimen Collection Kits, Therapak®
Supplier: Therapak, LLC
40003 40004 40005 40010
89171-052CS 209.55 CAD
89171-052 89428-908 10029-338 10029-340 10029-342 10029-344
Single Specimen Collection Kits, Therapak®
Sample Containers General Purpose Containers
Single Specimen Collection Kits can be used in conjunction with on-site testing products or remote site collection when specimen is transported to a laboratory for analysis.
Each kit contains a leak-resistant 90 ml sterile container with cap and integrity seal. The transport bag features two sealable, tamper-evident pockets, and comes with absorbent material. The individually bagged kits are available in various configurations.
Order Now
SPECIFICATIONS
Environmentally Preferable