Silver nitrate 0.10 N, Supelco®

Supplier: MilliporeSigma

SX0206A-2 SX0206A-5
CASX0206A-2CS 1306 CAD
CASX0206A-2 CASX0206A-5
Silver nitrate 0.10 N, Supelco®
Silver nitrate

Silver nitrate solution (0.10N).

Formula: AgNO₃
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 1493
ADR: 5.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Chemical formula AgNO₃ in water
Molar Mass 169.87
Key Applications General Chemistry
Color limit 10 max.
Traceability NIST Traceable
Expiration date Exp. 18 months from mfg. date
Appearance Clear liquid
Color (APHA) 10 max
Concentration (normality) 0.0999-0.1001N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR