Formalin 20% in aqueous solution, phosphate buffered, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

M-Clarity™ quality level = MQ300

R04587-01 R04587-86
CAR04587-01EA 297 CAD
CAR04587-01 CAR04587-86
Formalin 20% in aqueous solution, phosphate buffered, Sigma-Aldrich®
Formaldehyde

Formalin 20%, Phosphate Buffered

Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: -19.3
Melting Pt: -92
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 3334
Merck Index: 13,04259

Order Now

Specification Test Results

N/A N/A


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR