Chlorhexidine, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.43969.0050 8.43969.0250
CA8.43969.0050EA 231 CAD
CA8.43969.0050 CA8.43969.0250
Chlorhexidine, Sigma-Aldrich®
Chlorhexidine

Chlorohexidine for synthesis

Formula: C₂₂H₃₀Cl₂N₁₀
MDL Number: MFCD00009673
CAS Number: 55-56-1
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Hill Formula C₂₂H₃₀Cl₂N₁₀
Molar Mass 505.45 g/mol
Melting Point 134 - 136 °C
Solubility 0.8 g/l (20 °C)
LD 50 oral LD50 Rat 5000 mg/kg
Storage class 10 - 13 Other liquids and solids
WGK WGK 3 highly water endangering
Categories of danger irritant, dangerous for the environment
R Phrase R 41-51/53
S Phrase S 26-39-61


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR