Sodium molybdate(VI) dihydrate ≥98.0% (by titration analysis on dried basis) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Molybdic acid, sodium salt dihydrate, Molybdic acid sodium salt dihydrate, Molybdic acid disodium salt dihydrate, di-Sodium molybdate dihydrate, Sodium molybdate dihydrate, Sodium molybdenum oxide dihydrate

M-Clarity™ quality level = MQ200

8.43899.0050 8.43899.0250
CA8.43899.0050EA 108 CAD
CA8.43899.0050 CA8.43899.0250
Sodium molybdate(VI) dihydrate ≥98.0% (by titration analysis on dried basis) for synthesis, Sigma-Aldrich®
Sodium molybdate(VI) dihydrate

Sodium molybdate dihydrate for synthesis

Formula: Na₂MoO₄·2H₂O
MW: 241.95 g/mol
Melting Pt: 687
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
Merck Index: 13,08718

Order Now

Specification Test Results

Hill Formula Na₂MoO₄ * 2 H₂O
Chemical formula Na₂MoO₄ * 2 H₂O
Molar Mass 241.95 g/mol
pH value 9 - 10 (840 g/l, H₂O, 20 °C)
Bulk density 1000 - 1400 kg/m³
Solubility 840 g/l (20 °C)
LD 50 oral LD50 Rat 4233 mg/kg
LD 50 dermal LD50 Rat 2000 mg/kg
Storage class 10 - 13 Other liquids and solids
WGK WGK 1 slightly water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR