4-Nitroaniline, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: p-Nitroaniline, pNA

M-Clarity™ quality level = MQ200

8.22292.0250 8.22292.1000 8.22292.0250 8.22292.1000
EM8.22292.0250CS 477.16 CAD
EM8.22292.0250 EM8.22292.1000 CA8.22292.0250 CA8.22292.1000
4-Nitroaniline, Sigma-Aldrich®
4-Nitroaniline

4-Nitroaniline for synthesis

Formula: C₆H₆N₂O₂
MDL Number: MFCD00007858
CAS Number: 100-01-6
UN: 1661
ADR: 6.1,II

Order Now

Specification Test Results

Hill Formula C₆H₆N₂O₂
Chemical formula H₂NC₆H₄NO₂
Molar Mass 138.13 g/mol
Boiling point 331 °C (1013 hPa) (decomposition)
Density 1.44 g/cm³ (20 °C)
Flash point 199 °C
Ignition temperature 500 °C
Melting Point 148 °C
pH value (0.5 g/l, H₂O, 20 °C) neutral
Vapor pressure 1.33 hPa (142 °C)
Bulk density 630 kg/m³
Solubility 0.5 g/l (20 °C)
Storage class 6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK WGK 3 highly water endangering
R Phrase R 23/24/25-33-52/53
S Phrase S 28-36/37-45-61


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR