Tetrabutylammonium iodide, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

Synonyms: Tetra-n-butylammonium iodide, TBAI

M-Clarity™ quality level = MQ200

8.22152.0100 8.22152.0250 8.22152.0100 8.22152.0250
EM8.22152.0100CS 1828.06 CAD
EM8.22152.0100 EM8.22152.0250 CA8.22152.0100 CA8.22152.0250
Tetrabutylammonium iodide, Sigma-Aldrich®
Tetrabutylammonium iodide

Tetra-n-butylammonium iodide for synthesis

Formula: C₁₆H₃₆IN
MDL Number: MFCD00011636
CAS Number: 311-28-4

Order Now

Specification Test Results

Hill Formula C₁₆H₃₆IN
Molar Mass 369.38 g/mol
Melting Point 143 - 146 °C
LD 50 oral LD50 Rat 1990 mg/kg
Storage class 10 - 13 Other liquids and solids
WGK WGK 3 highly water endangering
Categories of danger harmful
R Phrase R 22
S Phrase S 22-24/25


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR