N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: 1,1',1'',1'''-(Ethane-1,2-diylbis(azanetriyl))tetrakis(propan-2-ol), Ethylenediamine-N,N,N',N'-tetra-2-propanol

M-Clarity™ quality level = MQ200

8.21940.0100 8.21940.1000
CA8.21940.0100CS 872 CAD
CA8.21940.0100 CA8.21940.1000
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine, Sigma-Aldrich®
N,N,N',N'-Tetrakis(2-hydroxypropyl)ethylenediamine

Ethylenediamine-N,N,N',N'-tetra-2-propanol for synthesis

Formula: C₁₄H₃₂N₂O₄
MW: 292.42 g/mol
MDL Number: MFCD00004534
CAS Number: 102-60-3

Order Now

Specification Test Results

Hill Formula C₁₄H₃₂N₂O₄
Chemical formula [CH₃CH(OH)CH₂]₂NCH₂CH₂N[CH₂CH(OH)CH₃]₂
Molar Mass 292.41 g/mol
Boiling point 240 °C (13 hPa)
Density 1.03 g/cm³ (20 °C)
pH value 10.4 (10 g/l, H₂O, 20 °C)
Refractive Index 1.478 (20 °C, 589 nm)
Solubility (20 °C) soluble
LD 50 oral LD50 Rat 2000 mg/kg
Storage class 10 - 13 Other liquids and solids
WGK WGK 3 highly water endangering

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR