Aniline hydrochloride, Sigma-Aldrich®
Supplier: MilliporeSigma
|
Danger
|
M-Clarity™ quality level = MQ200
8.20099.0250
8.20099.1000
8.20099.0250
8.20099.1000
EM8.20099.0250CS
4221.76
CAD
EM8.20099.0250
EM8.20099.1000
CA8.20099.0250
CA8.20099.1000
Aniline hydrochloride, Sigma-Aldrich®
Aniline hydrochloride
Anilinium chloride for synthesis
Formula:
C₆H₈ClN |
MDL Number:
MFCD00012958 CAS Number: 142-04-1 UN: 1548 ADR: 6.1,III |
Specification Test Results
Hill Formula | C₆H₈ClN |
Chemical formula | C₆H₅NH₂ * HCl |
Molar Mass | 129.58 g/mol |
Boiling point | 245 °C (1013 hPa) |
Flash point | 193 °C |
Melting Point | 199 - 202 °C |
Bulk density | 500 kg/m³ |
Solubility | 1070 g/l (25 °C) |
LD 50 oral | LD50 Rat 840 mg/kg |
Storage class | 6.1A Combustible substances, toxic |
WGK | WGK 2 water endangering |
R Phrase | R 23/24/25-40-41-43-48/23/24/25-68-50 |
S Phrase | S 26-27-36/37/39-45-61-63 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...