Tributyltin hydride, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Tri-n-butyltin hydride

M-Clarity™ quality level = MQ200

8.14109.0005 8.14109.0025 8.14109.0100 8.14109.0005 8.14109.0025 8.14109.0100
EM8.14109.0005CS 86249.04 CAD
EM8.14109.0005 EM8.14109.0025 EM8.14109.0100 CA8.14109.0005 CA8.14109.0025 CA8.14109.0100
Tributyltin hydride, Sigma-Aldrich®
Tributyltin hydride

Tributyltin hydride for synthesis

Formula: C₁₂H₂₈Sn
MW: 291.06 g/mol
MDL Number: MFCD00009416
CAS Number: 688-73-3
UN: 2788
ADR: 6.1,II

Order Now

Specification Test Results

Hill Formula C₁₂H₂₈Sn
Molar Mass 291.06 g/mol
Boiling point 112 - 114 °C (11 hPa)
Density 1.10 g/cm³ (20 °C)
Flash point 109 °C
Ignition temperature 250 °C
Vapor pressure 5 hPa (20 °C)
Refractive Index 1.4726 (20 °C, 589 nm)
Storage class 6.1A Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK WGK 3 highly water endangering
R 21-25-36/38-48/23/25-50/53 S Phrase
S 35-36/37/39-45-60-61

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR