2,2,2-Trifluoroethanol, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: Trifluoro Ethanol, TFE

M-Clarity™ quality level = MQ200

8.08259.0100 8.08259.0250 8.08259.1000 8.08259.0100 8.08259.0250 8.08259.1000
EM8.08259.0100CS 1576.84 CAD
EM8.08259.0100 EM8.08259.0250 EM8.08259.1000 CA8.08259.0100 CA8.08259.0250 CA8.08259.1000
2,2,2-Trifluoroethanol, Sigma-Aldrich®
2,2,2-Trifluoroethanol

2,2,2-Trifluoroethanol for synthesis

Formula: C₂H₃F₃O
MW: 100.04 g/mol
Boiling Pt: 74.1
Melting Pt: -43.5
Flash Pt: 33
MDL Number: MFCD00004672
CAS Number: 75-89-8
UN: 1992
ADR: 3 (6.1) III

Order Now

Specification Test Results

Hill Formula C₂H₃F₃O
Chemical formula CF₃CH₂OH
Molar Mass 100.04 g/mol
Boiling point 73.6 °C (1013 hPa)
Density 1.38 g/cm³ (20 °C)
Explosion limit 8.4 - 28.8 %(V)
Flash point 33 °C
Melting Point -43.5 °C
pH value (H₂O) neutral
Vapor pressure 52 hPa (20 °C)
Solubility (20 °C) soluble
LD 50 oral LD50 Rat 240 mg/kg
LD 50 dermal LD50 Rat 1680 mg/kg
Storage class 3 Flammable liquids
WGK WGK 1 slightly water endangering
R Phrase R 10-20/21/22-38-41-48/20
S Phrase S 26-36/37/39


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR