1-Octadecanol, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.07680.0100 8.07680.1000 8.07680.0100 8.07680.1000
EM8.07680.0100CS 1069.66 CAD
EM8.07680.0100 EM8.07680.1000 CA8.07680.0100 CA8.07680.1000
1-Octadecanol, Sigma-Aldrich®
1-Octadecanol

Stearyl alcohol for synthesis

Formula: H₃C(CH₂)₁₆CH₂OH
MDL Number: MFCD00002823
CAS Number: 112-92-5

Order Now

Specification Test Results

Hill Formula C₁₈H₃₈O
Molar Mass 270.49 g/mol
Boiling point 330 - 360 °C (1013 hPa)
Density 0.805 - 0.815 g/cm³ (60 °C)
Flash point 195 °C
Ignition temperature 230 °C DIN 51794
Melting Point 55 - 60 °C
Vapor pressure  - 1 hPa (20 °C)
Bulk density 300 kg/m³
Solubility (20 °C) practically insoluble
LD 50 oral LD50 Rat 2000 mg/kg
Storage class 10 - 13 Other liquids and solids
WGK NWG not water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR