Ethyl laurate, Sigma-Aldrich®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

8.05334.0005 8.05334.0250
CA8.05334.0005EA 40 CAD
CA8.05334.0005 CA8.05334.0250
Ethyl laurate, Sigma-Aldrich®
Ethyl laurate

Ethyl laurate for synthesis

Formula: C₁₄H₂₈O₂
MDL Number: MFCD00015065
CAS Number: 106-33-2

Order Now

Specification Test Results

Hill Formula C₁₄H₂₈O₂
Chemical formula CH₃(CH₂)₁₀COOC₂H₅
Molar Mass 228.37 g/mol
Boiling point 272 °C (1013 hPa)
Density 0.86 g/cm³ (20 °C)
Flash point 138 °C
Ignition temperature 300 °C
Melting Point -2 °C
Vapor pressure 0.1 hPa (60 °C)
Refractive Index 1.4308 (20 °C, 589 nm)
Solubility (20 °C) insoluble
LD 50 oral LD50 Rat 5000 mg/kg
LD 50 dermal LD50 Rabbit 5000 mg/kg
Storage class 10 Combustible liquids not in Storage Class 3
WGK WGK 2 water endangering


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR