Furfuryl alcohol, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ200

8.04015.0100 8.04015.1000 8.04015.0100 8.04015.1000
EM8.04015.0100CS 1298.76 CAD
EM8.04015.0100 EM8.04015.1000 CA8.04015.0100 CA8.04015.1000
Furfuryl alcohol, Sigma-Aldrich®
Furfuryl alcohol

Furfuryl alcohol for synthesis

Formula: C₅H₆O₂
MDL Number: MFCD00003252
CAS Number: 98-00-0
UN: 2874
ADR: 6.1,III

Order Now

Specification Test Results

Hill Formula C₅H₆O₂
Molar Mass 98.1 g/mol
Boiling point 170 - 171 °C (1013 mbar)
Density 1.13 g/cm³ (20 °C)
Explosion limit 1.8 - 16.3 %(V)
Flash point 65 °C
Ignition temperature 390 °C
Melting Point -31 °C
pH value 6 (300 g/l, H₂O, 20 °C)
Vapor pressure 0.53 hPa (20 °C)
Refractive Index 1.4845 (20 °C, 589 nm)
Solubility (20 °C) soluble
LD 50 oral LD50 Rat 177 mg/kg
LD 50 dermal LD50 Rabbit 400 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR