1-Octanesulfonic acid sodium salt, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Sodium octane-1-sulfonate
, Sodium octane-1-sulphonate
M-Clarity™ quality level = MQ100
1.18307.0025
1.18307.0025
CA1.18307.0025EA
695.67
CAD
EM1.18307.0025
CA1.18307.0025
1-Octanesulfonic acid sodium salt, Supelco®
1-Octanesulphonic acid sodium salt
Octane-1-sulfonic acid sodium salt for ion pair chromatography LiChropur®.
Formula:
C₈H₁₇NaO₃S MW: 216.28 g/mol Melting Pt: 300 °C |
MDL Number:
MFCD00007544 CAS Number: 5324-84-5 |
Specification Test Results
Assay (alkalimetric, calculated on dried substance) | ≥ 99 % |
Identity (IR-spectrum) | conforms |
pH (100 g/l, water) | 5.5 - 7.5 |
UV-transmission (0.005 mol/l; 1 cm; Water) (at 200 nm) | ≥ 70 % |
UV-transmission (0.005 mol/l; 1 cm; Water) (at 220 nm) | ≥ 90 % |
UV-transmission (0.005 mol/l; 1 cm; Water) (at 250 nm) | ≥ 98 % |
Loss on drying (120 °C; 4h; Vacuum) | ≤ 2.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...