Histology Bucket with Lid, Therapak®
Supplier: Therapak
75100G 75098G 75099G
10497-264CS 268.98 CAD
10497-264 10497-268 10497-266
Histology Bucket with Lid, Therapak®
Buckets
These polypropylene histology buckets are conveniently packed in case quantities and each bucket features a carrying handle and snap-on lid
The lid secures tightly to the bucket to reduce odor. The buckets are autoclavable, easy to stack, and are resistant to cracking when exposed to common cytology and histology based chemicals such as formaldehyde.