VWR® Bulk Beads
Supplier: VWR International
10158-550 10158-552 10158-554
10158-550EA 557.91 CAD
10158-550 10158-552 10158-554
VWR® Bulk Beads
Beads Metallic Beads
Bead beating is the most effective and verstile lysis method available.
  • Certified free of RNase and DNase
  • Designed for use on all bead mill platforms
  • Effective and versatile
VWR® offers optimal flexibility with a range of bead beating lysing matrices to support efficient homogenization of a wide range of samples.
VWR® bulk lysing matrix bead media is certified RNase and DNase free to ensure high analyte integrity. Beads are designed for use on all bead mill platforms including the VWR® Mini Bead Mill.
Certifications: RNase- and DNase-free.