VWR® Pre-Filled Bead Tubes, 2 ml

Supplier: VWR International
19-620-VWR 19-622-VWR 19-627-VWR 19-628-VWR
10158-604PK 332.35 CAD
10158-604 10158-606 10158-610 10158-612
VWR® Pre-Filled Bead Tubes, 2 ml
Tubes Centrifuge Tubes
VWR® pre-filled bead tubes offer convenience and flexibility with a range of bead beating tubes and lysing matrices to support efficient homogenization of a wide range of samples on most popular bead mill platforms.

  • Polypropylene fabrication
  • Pre-filled with an optimized amount of lysing matrix
  • Provide efficient, consistent, and high-yield sample homogenates

These VWR® bead beating tubes are pre-filled with an optimized amount of lysing matrix to provide efficient, consistent, and high-yield sample homogenates

These 2 ml polypropylene screw capMore Product Information
Order Now

SPECIFICATIONS

Operating temperature –80 to 121°C (–112 to 250°F)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR