VWR® Pre-Filled Bead Tubes, 1.5 ml
Supplier: VWR
19-610-VWR 19-617-VWR 19-618-VWR
10158-600PK 332.32 CAD
10158-600 10158-598 10158-602
VWR® Pre-Filled Bead Tubes, 1.5 ml
Tubes Centrifuge Tubes
VWR® pre-filled bead tubes offer convenience and flexibility with a range of bead beating tubes and lysing matrices to support efficient homogenization of a wide range of samples on most popular bead mill platforms.
  • Certified RNase and DNase free to ensure high analyte integrity
The VWR® 1.5 ml pre-filled bead tubes are compatible with the VWR® 4-Place Mini Bead Mill, Omni International Bead Ruptor 12, and Next Advance Bullet Blender.
These 1.5 ml polypropylene microtubes feature an attached lid and retaining cam. They are pre-filled with lysing matrix. Bead beating tubes are designed to withstand the high forces generated through bead milling.
These VWR® bead beating tubes are pre-filled with an optimized amount of lysing matrix to provide efficient, consistent, and high yield sample homogenates.
Ordering information: Unit includes 50 bead beating tubes pre-filled with lysing matrix.