beta-N-Acetylglucosaminidase S
Supplier: New England Biolabs (NEB)
P0744L
P0744S
P0744L
P0744S
102877-624EA
1000.14
CAD
102877-624
102877-626
CA102877-624
CA102877-626
beta-N-Acetylglucosaminidase S
Enzymes
β-N-Acetylglucosaminidase S is a highly specific exoglycosidase that catalyzes the hydrolysis of terminal, non-reducing β-N-Acetylglucosamine residues from oligosaccharides.