Alpha 2-3 Neuraminidase S
Supplier: New England Biolabs (NEB)
P0743L
P0743S
P0743L
P0743S
102877-620EA
522.98
CAD
102877-620
102877-622
CA102877-620
CA102877-622
Alpha 2-3 Neuraminidase S
Enzymes
Neuraminidase is the common name for Acetyl-neuraminyl hydrolase (Sialidase)
α2-3 Neuraminidase S is a highly specific exoglycosidase that catalyzes the hydrolysis of α2-3 linked N-acetyl-neuraminic acid residues from glycoproteins and oligosaccharides.