Alpha 2-3,6,8,9 Neuraminidase A
Supplier: New England Biolabs (NEB)
P0722L P0722S P0722L P0722S
102877-616EA 527.72 CAD
102877-616 102877-618 CA102877-616 CA102877-618
Alpha 2-3,6,8,9 Neuraminidase A
Enzymes
Neuraminidase is the common name for Acetyl-neuraminyl hydrolase (Sialidase)
α2-3,6,8,9 Neuraminidase A catalyzes the hydrolysis of all linear and branched non-reducing terminal sialic acid residues from glycoproteins and oligosaccharides. The enzyme releases α2-3 and α2-6 linkages at a slightly higher rate than α2-8 and α2-9 linkages.