Recombinant PNGase F
Supplier: New England Biolabs (NEB)
P0708L P0708S P0708L P0708S
102877-602EA 1215.02 CAD
102877-602 102877-604 CA102877-602 CA102877-604
Recombinant PNGase F
Enzymes
Peptide-N-Glycosidase F, also known as PNGase F, is a recombinant amidase which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides from N-linked glycoproteins.