S. pyogenes Cas9 Nuclease
Supplier: New England Biolabs (NEB)
M0386S
M0386M
M0386T
M0386S
M0386M
M0386T
102877-594EA
86.9
CAD
102877-594
102902-446
103218-912
CA102877-594
CA102902-446
CA103218-912
S. pyogenes Cas9 Nuclease
Enzymes
Cas9 Nuclease, S. pyogenes.
- Ideal for in vitro digestion of dsDNA
- Compatible with EnGen sgRNA Synthesis Kit, S. pyogenes and the EnGen Mutation Detection Kit
- For help with oligo design, try EnGen sgRNA Template Oligo Designer
Cas9 Nuclease, S. pyogenes, is an RNA-guided endonuclease that catalyzes site-specific cleavage of double stranded DNA. The location of the break is within the target sequence 3 bases from the NGG PAM (Protospacer Adjacent Motif) . The PAM sequence, NGG, must follow the targeted region on the opposite strand of the DNA with respect to the region complementary sgRNA sequence.