QUICK-LOAD® Purple DNA Ladder, New England BioLabs
Supplier: New England Biolabs (NEB)
N0551S N0552S N0551S N0552S N0550L N0550S N0551L N0552L
102877-810EA 159.58 CAD
102877-810 102877-812 CA102877-810 CA102877-812 CA103258-568 CA102877-598 76199-128 76199-130
QUICK-LOAD® Purple DNA Ladder, New England BioLabs
Electrophoresis Reagents Molecular Weight Markers
Quick-Load Purple 2-Log DNA Ladder is a pre-mixed, ready-to-load molecular weight marker containing one tracking dye
Tracking dye does not leave a shadow under UV light. Quick-Load Purple 2-Log DNA Ladder consists of proprietary plasmids, which are digested to completion with appropriate restriction enzymes to yield 19 bands suitable for use as molecular weight standards for agarose gel electrophoresis. The digested DNA includes fragments ranging from 100 bp to 10 kb. The 0.5, 1.0 and 3.0 kb bands have increased intensity to serve as reference bands.
Packaging: The Quick-Load Purple DNA Ladders are supplied with Gel Loading Dye, Purple (6X). Not supplied with SDS.