E.coli RNA Polymerase, Core Enzyme, New England Biolabs
Supplier: New England Biolabs
M0550S M0550S
CA102855-166EA 305 CAD
102855-166 CA102855-166
E.coli RNA Polymerase, Core Enzyme, New England Biolabs
Nucleic Acid Reagents RNA Synthesis and Transcription
E.coli RNA Polymerase Core Enzyme consists of 5 subunits designated a, a, b´, β, and ω. The enzyme is free of sigma factor and does not initiate specific transcription from bacterial and phage DNA promoters. The enzyme retains the ability to transcribe RNA from nonspecific initiation sequences. Addition of sigma factors will allow the enzyme to initiate RNA synthesis from specific bacterial and phage promoters. The core enzyme has a molecular weight of approximately 400 kDa.