Exonuclease VII
Supplier: New England Biolabs (NEB)
M0379L
M0379S
M0379L
M0379S
102877-588EA
1191.32
CAD
102877-588
102877-590
CA102877-588
CA102877-590
Exonuclease VII
Enzymes
Exonuclease VII, (Exo VII) derived from E.coli, cleaves single-stranded DNA (ssDNA) from both 5´→3´ and 3´→5´ direction. This enzyme is not active on linear or circular dsDNA.
It is useful for removal of single stranded oligonucleotide primers from a completed PCR reaction when different primers are required for subsequent PCR reactions. Digestion of ssDNA by Exonuclease VII is metal-independent.