NEBNEXT® Multiplex Small RNA Library Prep Set for Illumina (Set 2), New England Biolabs
Supplier: New England Biolabs (NEB)
E7580L E7580S E7580L E7580S
102877-576EA 8799.02 CAD
102877-576 102877-578 CA102877-576 CA102877-578
NEBNEXT® Multiplex Small RNA Library Prep Set for Illumina (Set 2), New England Biolabs
Nucleic Acid Reagents Next Generation Sequencing Reagents
The novel NEBNext® Small RNA workflow has been optimized to minimize adaptor-dimers while producing high-yield, high-diversity libraries
The NEBNext® Small RNA Library Prep Set for Illumina® (Set 2) includes adaptors and multiplex primers with 12 indices, to enable multiplexing.