NEBNEXT® ULTRA™ Directional RNA Second Strand Synthesis Module, New England Biolabs
Supplier: New England Biolabs (NEB)
E7550S E7550L
CA102855-116EA 610 CAD
CA102855-116 CA102855-132
NEBNEXT® ULTRA™ Directional RNA Second Strand Synthesis Module, New England Biolabs
Nucleic Acid Reagents RT-PCR and cDNA Reagents
The NEBNext® Ultra™ Directional RNA Second Strand Synthesis Module has been optimized to generate double-stranded cDNA from first-strand cDNA
The dsDNA generated by the NEBNext Ultra Directional RNA Second Strand Module can be subsequently converted to blunt-end DNA fragments using the NEBNext Ultra End Prep Module.