NEBUILDER HiFi DNA Assembly® Master Mix, New England Biolabs
Supplier: New England Biolabs (NEB)
E2621X E2621L E2621S E2621X
102877-806EA 4234.4 CAD
102877-806 CA102877-802 CA102877-804 CA102877-806
NEBUILDER HiFi DNA Assembly® Master Mix, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
NEBuilder HiFi DNA Assembly Master Mix was developed to improve the efficiency and accuracy of DNA assembly.
This method allows for seamless assembly of multiple DNA fragments, regardless of fragment length or end compatibility. This method has been used to assemble either single-stranded oligonucleotides or different sizes of DNA fragments with varied overlaps (15–80 bp). It has utility for the synthetic biology community, as well as those interested in one-step cloning of multiple fragments due to its ease of use, flexibility and simple master-mix format. The reaction includes different enzymes that work together in the same buffer. The exonuclease creates single-stranded 3´ overhangs that facilitate the annealing of fragments that share complementarity at one end (the overlap region). The polymerase fills in gaps within each annealed fragment. The DNA ligase seals nicks in the assembled DNA. The end result is a double-stranded fully sealed DNA molecule that can serve as template for PCR, RCA or a variety of other molecular biology applications, including direct transformation of E coli.