HISCRIBE™ T7 Quick High Yield RNA Synthesis Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E2050S E2050L
CA102855-162EA 466 CAD
CA102855-162 CA77532-236
HISCRIBE™ T7 Quick High Yield RNA Synthesis Kit, New England Biolabs
Nucleic Acid Reagents RNA Synthesis and Transcription
The HiScribe T7 Quick High Yield RNA Synthesis Kit offers robust in vitro RNA transcription for a variety of applications
The master mix format allows for quick reaction setup. DNase I and LiCl are included for DNA template removal and quick RNA purification. In addition, the kit allows for capped RNA or dye labeled RNA synthesis by incorporation of cap analog (ARCA) or dye modified nucleotides