NEB® PCR Cloning Kit, New England Biolabs
Supplier: New England Biolabs (NEB)
E1202S E1202S
102877-564EA 532.46 CAD
102877-564 CA102877-564
NEB® PCR Cloning Kit, New England Biolabs
Nucleic Acid Reagents Cloning Kits and Ligation
This PCR Cloning Kit contains an optimized 2X Cloning Master Mix with a proprietary ligation enhancer and a linearized vector that uses a novel mechanism for background colony suppression to give a low background.
It allows simple and quick cloning of any PCR amplicon, whether the amplification reactions are performed with proofreading DNA polymerases, such as Q5® or Phusion® which produce blunt ends; or nonproofreading DNA polymerases, such as Taq or Taq mixes (OneTaq®, LongAmp™ Taq) which produce single base overhangs. This is possible due to “invisible” end polishing components in the master mix that are active during the ligation step. The kit also allows direct cloning from amplification reactions without purification, and works well whether or not the primers used in the PCR possess 5´-phosphate groups.