Gel Loading Dye Purple (6X), New England Biolabs
Supplier: New England Biolabs (NEB)
B7024S B7024S
102877-610EA 82.16 CAD
102877-610 CA102877-610
Gel Loading Dye Purple (6X), New England Biolabs
Stains Electrophoresis Stains
Gel Loading Dye, Purple (6X) is a pre-mixed loading buffer which contains a combination of two dyes, Dye 1 (pink/red) and Dye 2 (blue).
The red dye serves as the tracking dye for both agarose and non-denaturing polyacrylamide gel electrophoresis. The two dyes separate upon gel electrophoresis; the red band is the major indicator and migrates similarly to Bromophenol Blue on agarose gels. Specifically chosen, this dye does not leave a shadow under UV light. This solution contains SDS, which often results in sharper bands, as some restriction enzymes are known to remain bound to DNA following cleavage. EDTA is also included to chelate magnesium (up to 10 mM) in enzymatic reactions, thereby stopping the reaction. The dye also contains Ficoll®, which creates brighter and tighter bands when compared to glycerol loading dyes.