Remove-iT Endo D
Supplier: New England Biolabs (NEB)
P0742S
P0742L
CA102855-184EA
336
CAD
CA102855-184
CA102855-186
Remove-iT Endo D
Enzymes
Remove-iT® Endo D, also known as Endoglycosidase D, is a recombinant glycosidase, which cleaves within the chitobiose core of paucimannose N-linked glycans, with or without extensions in the antennae
Remove-iT Endo D is tagged with a chitin binding domain (CBD) for easy removal from a reaction, and is supplied glycerol-free for optimal performance in HPLC and MS intensive methods.