Remove-iT Endo S
Supplier: New England Biolabs (NEB)
P0741S
P0741L
CA102855-180EA
336
CAD
CA102855-180
CA102855-182
Remove-iT Endo S
Enzymes
Remove-iT® Endo S is an endoglycosidase with a uniquely high specificity for removing N-linked glycans from the chitobiose core of the heavy chain of native IgG
Remove-iT Endo S is tagged with a chitin binding domain (CBD) for easy removal from a reaction and is supplied glycerol free for optimal performance in HPLC and MS intensive methods.