SGE Syringes, GC Autosampler Syringe, CTC, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
001981 002985 005335 002980 002981 003987 002992 002982 002984 001982 003988 005333 002986 002987 006700 001984 002700 002967 002971 002972 002976 002977 002983 003700 005700 0039871
14225-174EA 287.99 CAD
14225-174 14225-234 14225-318 60361-140 60361-142 60361-156 60361-364 60361-380 60361-382 60362-182 60362-654 60362-660 60362-676 60361-144 89209-054 89209-082 89209-084 89209-088 89209-090 89209-092 89209-094 89209-096 89209-098 89209-100 89209-104 10799-826
SGE Syringes, GC Autosampler Syringe, CTC, Trajan Scientific and Medical
Syringes Syringes for Chromatography
SGE has been making precision chromatography syringes for over 40 years
  • Unique plunger design minimizes bending and seizing
  • Cone tip needle point style has been developed specifically to withstand multiple fast septum injections
Metal plungers are individually fitted to the syringe glass barrel for a perfect “feel”, optimized life with minimal carry over, a liquid tight seal between the barrel and plunger and excellent performance.
PTFE tipped replaceable plunger syringes reduce the risk of plunger seizing when working with dirty samples, reducingMore Product Information