TruLine ORP 50 Electrode, YSI
Supplier: YSI
TruLine
400355
10122-562EA 608 CAD
10122-562
TruLine ORP 50 Electrode, YSI
Electrodes Redox Electrodes
TruLine ORP 50, ORP electrode, Single Junction, Refillable, Glass shaft, 1 m cable, BNC Connector.
Ag/AgCl Reference System. SMEK plug head.