VWR® Lanced and Unlanced Cylindrical Closures, Stainless Steel
Supplier: VWR International
10200-136 10200-138 10200-140 10200-142 10200-144 10204-066 10204-068 10204-070 10204-072 10204-074
10200-136CS 3360.19 CAD
10200-136 10200-138 10200-140 10200-142 10200-144 10204-066 10204-068 10204-070 10204-072 10204-074
VWR® Lanced and Unlanced Cylindrical Closures, Stainless Steel
Closures Test/Sample Tube Closures
Non-contaminating, noncorrosive stainless steel cylindrical closures are available in lanced and unlanced formats.
  • Prevents dust and microorganism buildup
  • Reduce amount of specimen evaporation by 50% compared to cotton plugging
  • Withstand repeated autoclaving without loss of resiliency
  • Available in plain or lanced formats
  • Lanced closures feature three resilient fingers that Adjust to nominal variations in tube O.D.
Top surface indentations allow adequate gas exchange under aerobic and anaerobic conditions.
Order Now
SPECIFICATIONS
Environmentally Preferable