1,3-Dimethyl-2-imidazolidinone ≥99.0% (by GC) for synthesis, Sigma-Aldrich®

Supplier: MilliporeSigma
Danger

Synonyms: 1,3-Dimethylimidazolidin-2-one

M-Clarity™ quality level = MQ200

8.18212.0500 8.18212.6190
EM8.18212.0500CS 2791.86 CAD
EM8.18212.0500 EM8.18212.6190
1,3-Dimethyl-2-imidazolidinone ≥99.0% (by GC) for synthesis, Sigma-Aldrich®
1,3-Dimethyl-2-imidazolidinone
Formula: C₅H₁₀N₂O
MW: 114.15 g/mol
Boiling Pt: 225.5
Melting Pt: 8
Flash Pt: 93
MDL Number: MFCD00003188
CAS Number: 80-73-9
UN: 2810
Merck Index: 13,03275

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 8.2 °C
Molar Mass 114.14 g/mol
Boiling Point 104 °C (20 hPa)
Vapor Pressure  - 1 hPa (20 °C)
Flash Point 114 °C
Refractive Index 1.472 (20 °C, 589 nm)
Explosion Limit 1.3 - 8.4 %(V)
Density 1.056 g/cm3 (20 °C)
pH (H2O) neutral
Ignition Point 305 °C
Synonyms Dimethylethylene urea, DMI, DMEU
Hill Formula C5H10N2O


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR