Alkali blue (C.I. 42765) solution indicator, Supelco®

Supplier: MilliporeSigma
Danger

1.09198.1000 1.09198.2500
CA1.09198.1000EA 334 CAD
CA1.09198.1000 CA1.09198.2500
Alkali blue (C.I. 42765) solution indicator, Supelco®
Alkali blue (C.I. 42765)
Formula: C₃₇H₂₉N₃O₃S
MW: 595.73 g/mol
Boiling Pt: 80 °C (1013 hPa)
Density: 0.84 g/cm3 (20 °C)
Flash Pt: 4 °C
Storage Temperature: Ambient
MDL Number: MFCD00083221
CAS Number: 1324-76-1
UN: 1993
ADR: 3 II

Order Now

Specification Test Results

dentity (UV/VIS-Spectrum) passes test
Density (d 20 °C/ 4 °C) 0.836 - 0.838
Absorption maximum λmax. (1 : 10 diluted) 603 - 607 nm
Absorbance A (λmax; 1 : 10 diluted; 1 cm) ≥ 1.00
Suitability as indicator (according to DIN 51558) passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR