Schiff reagent Histology, Sigma-Aldrich®

Supplier: MilliporeSigma
Warning

1.09033.0500 1.09033.2500
CA1.09033.0500EA 323 CAD
CA1.09033.0500 CA1.09033.2500
Schiff reagent Histology, Sigma-Aldrich®
Schiff reagent

Ready-to-use staining solution for human-medical cell diagnosis.


Schiff‘s reagent for the periodic acid Schiff, or PAS reaction, is used for the detection of mucopolysaccharides, glycogen, muco- and glycoproteins, glyco-, phospholipids, basal membranes and collagen. With Schiff's reagent aldehydes give a bright red color reaction, thus in combination with the periodic acid the nuclei appear blue. 500 ml are sufficient for approx. 2500 - 3000 stainings. It is registered as IVD and CE certified product and can be used for clinical diagnostics.

Density: 1.01 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00081985
UN: 1789
ADR: 8 II
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR