Accessories for SGE Syringes, RSH Syringe, GC Autosampler, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
SGE Syringes
032815 032835 032831 037875 034780 032846 032821 036871 037871 032810 037877 032841 037873 036875 036877 032825 036873
89236-306EA 74.43 CAD
89236-306 89236-328 89236-308 89236-320 89236-474 89236-310 89236-476 89236-312 89236-466 89236-478 89236-314 89236-468 89236-304 89236-316 89236-480 89236-470 89236-472
Accessories for SGE Syringes, RSH Syringe, GC Autosampler, Trajan Scientific and Medical
Syringes Syringes for Chromatography
Replacement Needles and Plungers for Thermo Scientific TriPlus RSH and CTC Analytics RTC Syringes.
  • Built to exacting standards
  • Deliver performance accuracy and precision
Ideal for a vast range of applications.