SGE Syringes, RSH Syringe, GC Autosampler, Trajan Scientific and Medical
Supplier: SGE (Trajan)
004864 002863 002862 007890 002869 002865 008655 001877 002872 002868 006868 003866 004868 005866 000680 007866 004862 001871 002875 006891 002861 007868 006864 002871 002878 004866 007891 001875 007864 001861 006890 007862 002864 003862 005891 003868 005864 006862 001863 005862 006866 005868 001865 005890 002866 003864 002876 000480 008155 002874 002867
10799-788EA 220.41 CAD
10799-788 89236-190 89236-236 89236-238 89236-192 89236-270 89236-272 89236-196 89236-152 89236-274 89236-154 89236-144 89236-188 89236-222 89236-266 89236-224 89236-268 89236-148 89236-226 89236-228 89236-180 89236-182 89236-260 89236-184 89236-140 89236-142 89236-264 89236-208 89236-254 89236-210 89236-178 89236-256 89236-212 89236-136 89236-258 89236-214 89236-138 89236-290 89236-170 89236-172 89236-250 89236-174 89236-252 89236-176 89236-166 89236-244 89236-168 89236-246 89236-240 89236-284 89236-242
SGE Syringes, RSH Syringe, GC Autosampler, Trajan Scientific and Medical
Syringes Syringes for Chromatography
SGE has been making precision chromatography syringes for over 40 years
Features and Benefits–Unique plunger design minimizes bending and seizing, and the cone tip needle point style has been developed specifically to withstand multiple fast septum injections.
Metal plungers are individually fitted to the syringe glass barrel for a perfect “feel”, optimized life with minimal carry over, a liquid tight seal between the barrel and plunger and excellent performance.
PTFE tipped replaceable plunger syringes reduce the risk of plunger seizing when working with dirty samples, reducing instrument downtime, and minimizing the risk of instrument shutdown during the processing of large batches of samples.
For NanoVolume syringes, the accuracy and reproducibility is ± 2 % (displaced volume); temperature range is 5-70°C. For Fixed Needle Syringes, accuracy and reproducibility is ± 1 % (displaced volume); temperature range is 5-70°C. For Removable Needle Syringes, accuracy and reproducibility is ± 1 % (displaced volume); temperature range is 5-120°C.
Our comprehensive range of GC autosampler syringes are designed and tested to meet critical autosampler specifications.
Certifications: RoHS compliant per Directive 2011/65/EU.
Ordering information: Wide range of fixed needle and removable needle options available. PTFE tipped replaceable plunger versions are available for use with gas and headspace samples.