Anti-KRAS Mouse Monoclonal Antibody [clone: 4H4G7]

Supplier: Proteintech
60309-1-IG-150UL
10155-892EA 709.42 CAD
10155-892
Anti-KRAS Mouse Monoclonal Antibody [clone: 4H4G7]
Antibodies
The KRAS antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human KRAS. This antibody recognizes human antigen. The KRAS antibody has been validated for the following applications: ELISA, WB, IHC, FC analysis.

WB 1 : 600

Type: Primary
Antigen: KRAS
Clonality: Monoclonal
Clone: 4H4G7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR