SGE Syringes, RSH Syringe, LC Autosampler, Trajan Scientific and Medical
Supplier: Trajan Scientific and Medical
002880 008150 008151 008950 008951
89236-198EA 245.64 CAD
89236-198 89236-280 89236-282 89236-294 89236-296
SGE Syringes, RSH Syringe, LC Autosampler, Trajan Scientific and Medical
Syringes Syringes for Chromatography
SGE has been making precision chromatography syringes for over 40 years.
  • Unique plunger design minimizes bending and seizing
  • Needles have a 90° square tip with rounded and polished edges
PTFE tipped replaceable plunger syringes reduce the risk of plunger seizing when working with dirty samples, reducing instrument downtime, and minimizing the risk of instrument shutdown during the processing of large batches of samples.
For fixed needle syringes the accuracy and reproducibility is ± 1% (displaced volume); temperature range is 5-70 °C. For removable needle syringes the accuracy and reproducibility is ± 1% (displaced volume); temperature range is 5-120 °C.
Our comprehensive range of HPLC autosampler syringes are designed and tested to meet critical autosampler specifications.
Certifications: RoHS compliant per Directive 2011/65/EU.
Ordering information: Range of fixed needle and removable needle options available.