StabilZyme® Noble Stabilizer, Surmodics
Supplier: Surmodics
SZ04-0050 SZ04-1000
102466-334EA 63.2 CAD
102466-334 102466-336
StabilZyme® Noble Stabilizer, Surmodics
Protein Linking and Labeling Systems
StabilZyme® HRP, StabilZyme® Protein-Free and StabilZyme® NOBLE stabilizers provide the assay developer a complete set of tools to stabilize different HRP conjugates for long term, in-solution stability.
  • Extended shelf life of kits
  • Elimination of protein interference and cross reactivity with the purpose of reducing erroneous results within immunoassay applications
The StabilZyme NOBLE Stabilizer (BSA-Free) is optimized to maintain the conformation of antibody/antigen conjugates, antibody-coated particles and unmodified proteins that are commonly used in immunoassays. The key benefits of the BSA-free formulation are the elimination of protein interference and cross reactivity with the purpose of reducing erroneous results within immunoassay applications.
Certifications: ISO 13485:2016 & 9001:2015 Certified