StabilZyme® Protein-Free Stabilizer, Surmodics
Supplier: Surmodics
SZPF-0050
SZPF-1000
102466-338EA
63.2
CAD
102466-338
102466-340
StabilZyme® Protein-Free Stabilizer, Surmodics
Protein Linking and Labeling Systems
StabilZyme® HRP, StabilZyme® Protein-Free and StabilZyme® NOBLE stabilizers provide the assay developer a complete set of tools to stabilize different HRP conjugates for long term, in-solution stability.
- Protein-Free formulation
- Extended shelf life of kits
- Elimination of protein interference and cross reactivity with the purpose of reducing erroneous results within immunoassay applications
StabilZyme® Protein-Free Stabilizer eliminates interference and cross-reactivity caused by proteins, delivering optimal performance for even the most sensitive immunoassays.
Certifications: ISO 13485:2016 & 9001:2015 Certified