GC Autosampler CTC Headspace Syringes, SGE
Supplier: Trajan Scientific and Medical
008131 008136 008631 008636
89236-276EA 610.92 CAD
89236-276 89236-278 89236-286 89236-288
GC Autosampler CTC Headspace Syringes, SGE
Syringes Syringes for Chromatography
Features and Benefits–SGE’s unique energized plunger tip design ensures a gas tight seal is consistently maintained providing outstanding performance (%RSD) with temperature cycling.
  • Outstanding reproducibility maintained during temperature cycling
  • Highest operating temperature
  • Save time and improve analysis workflow
The PTFE plunger tip in Diamond Headspace Syringes enables the use of a broader range of samples and solvents compared with polymers used in plungers of other high temperature syringes, this provides greater flexibility for your headspaceMore Product Information