HyLite® 47-409 Nitrile-Coated Gloves, Ansell
Supplier: Ansell Healthcare
205950 205951 205952 205953
80092-606CS 2688.62 CAD
80092-606 80092-608 80092-610 80092-612
HyLite® 47-409 Nitrile-Coated Gloves, Ansell
Gloves
A smart alternative that outperforms cotton, leather, and PVC gloves, HyLite® 47-409 gloves outwear cotton up to 12-to-1, lightweight leather 5-to-1, and PVC-coated gloves 3-to-1.
  • Exceptional resistance to snags, punctures, abrasions, and cuts
  • Excellent flexibility
  • Unique interlock knit liner and contoured design allows greater dexterity
Gloves feature a comfortable cotton lining with nitrile coating for exceptional snag, puncture, abrasion and cut resistance.
Use in the general handling, janitorial services, sanitation, metal fabrication,More Product Information